Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAGGTGACAACCTCTTCTTCATC[A/G]TTGTCAAAGGGGTTGCTGTTCTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MKRN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MKRN1 - makorin ring finger protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145125.1 | 1316 | Intron | NP_001138597.1 | |||
NM_001291663.1 | 1316 | Intron | NP_001278592.1 | |||
NM_013446.3 | 1316 | Silent Mutation | AAC,AAT | N,N 434 | NP_038474.2 | |
XM_011515996.1 | 1316 | Silent Mutation | AAC,AAT | N,N 391 | XP_011514298.1 | |
XM_011515997.2 | 1316 | Silent Mutation | AAC,AAT | N,N 370 | XP_011514299.1 | |
XM_011515998.1 | 1316 | Intron | XP_011514300.1 |