Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTTACGGTGCCATCCCTTGACTC[A/C]ACCTTTAGAACAACGGGTTTTTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP3A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP3A4 - cytochrome P450 family 3 subfamily A member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202855.2 | 1583 | Silent Mutation | GTG,GTT | V,V 492 | NP_001189784.1 | |
NM_017460.5 | 1583 | Silent Mutation | GTG,GTT | V,V 493 | NP_059488.2 | |
XM_017011780.1 | 1583 | Intron | XP_016867269.1 |