Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGAACTTTTAAATTGATATCTT[C/T]TATCAGTTCTGAAAAATTTCACATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609996 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL28A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL28A1 - collagen type XXVIII alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001037763.2 | Intron | NP_001032852.2 | ||||
XM_011515358.2 | Intron | XP_011513660.1 | ||||
XM_011515359.2 | Intron | XP_011513661.1 | ||||
XM_011515360.2 | Intron | XP_011513662.1 | ||||
XM_011515362.2 | Intron | XP_011513664.1 | ||||
XM_011515363.2 | Intron | XP_011513665.1 | ||||
XM_011515364.2 | Intron | XP_011513666.1 | ||||
XM_011515365.2 | Intron | XP_011513667.1 | ||||
XM_011515366.2 | Intron | XP_011513668.1 | ||||
XM_017012131.1 | Intron | XP_016867620.1 | ||||
XM_017012132.1 | Intron | XP_016867621.1 | ||||
XM_017012133.1 | Intron | XP_016867622.1 |