Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCGCCAATTTGCCGCTGAACAGCC[G/T]CCAGAAGGAGCTGTGCAAGAGGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606267 | ||||||||||||||||||||
Literature Links: |
WNT16 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WNT16 - Wnt family member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016087.2 | 421 | Missense Mutation | CGC,CTC | R,L 45 | NP_057171.2 | |
NM_057168.1 | 421 | Missense Mutation | CGC,CTC | R,L 55 | NP_476509.1 |