Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACTGGCAGGCCACCATCATGGGCC[C/T]GGTAGGTAGTAGCTGCTGAGCGCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
POLR2J4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
POLR2J4 - RNA polymerase II subunit J4, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBE2D4 - ubiquitin conjugating enzyme E2 D4 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015983.3 | 309 | Intron | NP_057067.1 | |||
XM_006715734.3 | 309 | Intron | XP_006715797.1 | |||
XM_011515420.2 | 309 | Missense Mutation | CCG,CTG | P,L 60 | XP_011513722.1 | |
XM_011515422.2 | 309 | Intron | XP_011513724.1 | |||
XM_011515423.2 | 309 | Intron | XP_011513725.1 | |||
XM_017012292.1 | 309 | Missense Mutation | CCG,CTG | P,L 60 | XP_016867781.1 | |
XM_017012293.1 | 309 | Missense Mutation | CCG,CTG | P,L 60 | XP_016867782.1 | |
XM_017012294.1 | 309 | Missense Mutation | CCG,CTG | P,L 60 | XP_016867783.1 | |
XM_017012295.1 | 309 | Intron | XP_016867784.1 |