Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGATGCTGCAAATCAGGCCCTT[C/G]TCAGGTTTCTCCCAAACTCCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL26 - C-C motif chemokine ligand 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006072.4 | 173 | UTR 5 | NP_006063.1 | |||
XM_017011671.1 | 173 | Missense Mutation | GAC,GAG | D,E 47 | XP_016867160.1 | |
XM_017011672.1 | 173 | UTR 5 | XP_016867161.1 |