Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAAGGAAACATACTTAGCATCG[C/G]CACCACAGTCCCATAGCTCGAATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
IFT22 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT22 - intraflagellar transport 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130820.2 | 384 | Intron | NP_001124292.1 | |||
NM_001130821.2 | 384 | UTR 5 | NP_001124293.1 | |||
NM_001130822.2 | 384 | UTR 5 | NP_001124294.1 | |||
NM_001287525.1 | 384 | UTR 5 | NP_001274454.1 | |||
NM_001287526.1 | 384 | Intron | NP_001274455.1 | |||
NM_022777.3 | 384 | Missense Mutation | GCC,GGC | A,G 66 | NP_073614.1 |
LOC101927746 - uncharacterized LOC101927746 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |