Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGGCCTCTCGGAGAAAATGGCC[C/G]TCAGCATCGTGGACTTCCGCCGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EEPD1 - endonuclease/exonuclease/phosphatase family domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030636.2 | 1193 | Missense Mutation | CTC,GTC | L,V 159 | NP_085139.2 | |
XM_017012657.1 | 1193 | Missense Mutation | CTC,GTC | L,V 159 | XP_016868146.1 | |
XM_017012658.1 | 1193 | Missense Mutation | CTC,GTC | L,V 159 | XP_016868147.1 |