Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTAATTCCTCCCAGTATTTGA[G/T]GTTCCTTTATTACTTATTTTCACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603502 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFRD1 - interferon related developmental regulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007245.2 | 102 | UTR 5 | NP_001007246.1 | |||
NM_001197079.1 | 102 | Intron | NP_001184008.1 | |||
NM_001197080.1 | 102 | UTR 5 | NP_001184009.1 | |||
NM_001550.3 | 102 | Intron | NP_001541.2 | |||
XM_011516142.2 | 102 | Intron | XP_011514444.1 | |||
XM_011516143.2 | 102 | Intron | XP_011514445.1 | |||
XM_017012151.1 | 102 | Intron | XP_016867640.1 |