Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAAAAATCAGCAAACTAAATAT[A/G]AAAGGGAACTATCTCAACCAGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609700 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RABGEF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RABGEF1 - RAB guanine nucleotide exchange factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287060.1 | Intron | NP_001273989.1 | ||||
NM_001287061.1 | Intron | NP_001273990.1 | ||||
NM_001287062.1 | Intron | NP_001273991.1 | ||||
NM_014504.2 | Intron | NP_055319.1 |