Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCCACCATGAGTGCTGAGCTTAA[C/T]GTGCCTATCGACCCCTCTGCTCCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610929 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ORAI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ORAI2 - ORAI calcium release-activated calcium modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126340.2 | 163 | Silent Mutation | AAC,AAT | N,N 6 | NP_001119812.1 | |
NM_001271818.1 | 163 | Silent Mutation | AAC,AAT | N,N 6 | NP_001258747.1 | |
NM_001271819.1 | 163 | Intron | NP_001258748.1 | |||
NM_032831.3 | 163 | Silent Mutation | AAC,AAT | N,N 6 | NP_116220.1 |