Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGAGAAGAGATGATGTAAACTATA[C/T]GAGCATCCTTATGCCAAGCTCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOT1L1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOT1L1 - glutamic-oxaloacetic transaminase 1-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152413.2 | 853 | Missense Mutation | CAT,CGT | H,R 127 | NP_689626.2 | |
XM_005273399.3 | 853 | Missense Mutation | CAT,CGT | H,R 127 | XP_005273456.1 | |
XM_006716285.3 | 853 | Missense Mutation | CAT,CGT | H,R 127 | XP_006716348.1 | |
XM_011544391.2 | 853 | UTR 5 | XP_011542693.1 |