Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGAGGCATCCTGCCATACCTC[A/G]TGCTCAAAGGAGTCATCAAAGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600582 MIM: 611292 | ||||||||||||||||||||
Literature Links: |
ASPH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASPH - aspartate beta-hydroxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164750.1 | 2227 | Silent Mutation | CAC,CAT | H,H 696 | NP_001158222.1 | |
NM_001164751.1 | 2227 | Intron | NP_001158223.1 | |||
NM_001164752.1 | 2227 | Intron | NP_001158224.1 | |||
NM_001164753.1 | 2227 | Intron | NP_001158225.1 | |||
NM_001164754.1 | 2227 | Intron | NP_001158226.1 | |||
NM_001164755.1 | 2227 | Intron | NP_001158227.1 | |||
NM_001164756.1 | 2227 | Intron | NP_001158228.1 | |||
NM_004318.3 | 2227 | Silent Mutation | CAC,CAT | H,H 725 | NP_004309.2 | |
NM_020164.4 | 2227 | Intron | NP_064549.1 | |||
NM_032466.3 | 2227 | Intron | NP_115855.1 | |||
NM_032467.3 | 2227 | Intron | NP_115856.1 | |||
NM_032468.4 | 2227 | Intron | NP_115857.1 | |||
XM_005251235.2 | 2227 | Silent Mutation | CAC,CAT | H,H 740 | XP_005251292.1 | |
XM_005251236.2 | 2227 | Silent Mutation | CAC,CAT | H,H 739 | XP_005251293.1 | |
XM_005251238.2 | 2227 | Silent Mutation | CAC,CAT | H,H 725 | XP_005251295.1 | |
XM_005251239.2 | 2227 | Silent Mutation | CAC,CAT | H,H 721 | XP_005251296.1 | |
XM_005251240.1 | 2227 | Silent Mutation | CAC,CAT | H,H 711 | XP_005251297.1 | |
XM_005251242.2 | 2227 | Silent Mutation | CAC,CAT | H,H 706 | XP_005251299.1 | |
XM_005251243.2 | 2227 | Silent Mutation | CAC,CAT | H,H 697 | XP_005251300.1 | |
XM_005251244.1 | 2227 | Silent Mutation | CAC,CAT | H,H 691 | XP_005251301.1 | |
XM_005251246.2 | 2227 | Silent Mutation | CAC,CAT | H,H 682 | XP_005251303.1 | |
XM_005251247.2 | 2227 | Silent Mutation | CAC,CAT | H,H 678 | XP_005251304.1 | |
XM_005251248.1 | 2227 | Silent Mutation | CAC,CAT | H,H 677 | XP_005251305.1 | |
XM_005251250.2 | 2227 | Silent Mutation | CAC,CAT | H,H 663 | XP_005251307.1 | |
XM_017013419.1 | 2227 | Silent Mutation | CAC,CAT | H,H 767 | XP_016868908.1 | |
XM_017013420.1 | 2227 | Silent Mutation | CAC,CAT | H,H 766 | XP_016868909.1 | |
XM_017013421.1 | 2227 | Silent Mutation | CAC,CAT | H,H 753 | XP_016868910.1 | |
XM_017013422.1 | 2227 | Silent Mutation | CAC,CAT | H,H 752 | XP_016868911.1 | |
XM_017013423.1 | 2227 | Silent Mutation | CAC,CAT | H,H 752 | XP_016868912.1 | |
XM_017013424.1 | 2227 | Silent Mutation | CAC,CAT | H,H 751 | XP_016868913.1 | |
XM_017013425.1 | 2227 | Silent Mutation | CAC,CAT | H,H 748 | XP_016868914.1 | |
XM_017013426.1 | 2227 | Silent Mutation | CAC,CAT | H,H 738 | XP_016868915.1 | |
XM_017013427.1 | 2227 | Silent Mutation | CAC,CAT | H,H 737 | XP_016868916.1 | |
XM_017013428.1 | 2227 | Silent Mutation | CAC,CAT | H,H 733 | XP_016868917.1 | |
XM_017013429.1 | 2227 | Silent Mutation | CAC,CAT | H,H 724 | XP_016868918.1 | |
XM_017013430.1 | 2227 | Silent Mutation | CAC,CAT | H,H 724 | XP_016868919.1 | |
XM_017013431.1 | 2227 | Silent Mutation | CAC,CAT | H,H 723 | XP_016868920.1 | |
XM_017013432.1 | 2227 | Silent Mutation | CAC,CAT | H,H 723 | XP_016868921.1 | |
XM_017013433.1 | 2227 | Silent Mutation | CAC,CAT | H,H 722 | XP_016868922.1 | |
XM_017013434.1 | 2227 | Silent Mutation | CAC,CAT | H,H 720 | XP_016868923.1 | |
XM_017013435.1 | 2227 | Silent Mutation | CAC,CAT | H,H 718 | XP_016868924.1 | |
XM_017013436.1 | 2227 | Silent Mutation | CAC,CAT | H,H 711 | XP_016868925.1 | |
XM_017013437.1 | 2227 | Silent Mutation | CAC,CAT | H,H 709 | XP_016868926.1 | |
XM_017013438.1 | 2227 | Silent Mutation | CAC,CAT | H,H 696 | XP_016868927.1 | |
XM_017013439.1 | 2227 | Silent Mutation | CAC,CAT | H,H 695 | XP_016868928.1 | |
XM_017013440.1 | 2227 | Silent Mutation | CAC,CAT | H,H 695 | XP_016868929.1 | |
XM_017013441.1 | 2227 | Silent Mutation | CAC,CAT | H,H 681 | XP_016868930.1 | |
XM_017013442.1 | 2227 | Silent Mutation | CAC,CAT | H,H 677 | XP_016868931.1 | |
XM_017013443.1 | 2227 | Silent Mutation | CAC,CAT | H,H 662 | XP_016868932.1 | |
XM_017013444.1 | 2227 | Silent Mutation | CAC,CAT | H,H 653 | XP_016868933.1 | |
XM_017013445.1 | 2227 | Intron | XP_016868934.1 | |||
XM_017013446.1 | 2227 | Intron | XP_016868935.1 | |||
XM_017013447.1 | 2227 | Intron | XP_016868936.1 |
CLVS1 - clavesin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |