Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGTTCCGATGTGGTCCCCGGAG[A/C]GGGAGGCCGAGGCCCCAGCCGGGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611130 | ||||||||||||||||||||
Literature Links: |
CHMP7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP7 - charged multivesicular body protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317899.1 | 97 | UTR 5 | NP_001304828.1 | |||
NM_152272.4 | 97 | Silent Mutation | AGG,CGG | R,R 6 | NP_689485.1 | |
XM_006716415.2 | 97 | Silent Mutation | AGG,CGG | R,R 6 | XP_006716478.1 | |
XM_017013961.1 | 97 | Silent Mutation | AGG,CGG | R,R 6 | XP_016869450.1 | |
XM_017013962.1 | 97 | UTR 5 | XP_016869451.1 | |||
XM_017013963.1 | 97 | UTR 5 | XP_016869452.1 | |||
XM_017013964.1 | 97 | UTR 5 | XP_016869453.1 | |||
XM_017013965.1 | 97 | UTR 5 | XP_016869454.1 |