Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCGGCCCCGCGTGGCCGTGAAGAT[G/T]GCCTTCCGCAAGGCCTACTCCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130592 MIM: 616408 | ||||||||||||||||||||
Literature Links: |
EEF1D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEF1D - eukaryotic translation elongation factor 1 delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005250820.3 | 150 | Intron | XP_005250877.1 | |||
XM_005250823.1 | 150 | Intron | XP_005250880.1 | |||
XM_006716519.3 | 150 | Intron | XP_006716582.1 | |||
XM_006716520.3 | 150 | Intron | XP_006716583.1 | |||
XM_006716522.1 | 150 | Intron | XP_006716585.1 | |||
XM_006716523.1 | 150 | Intron | XP_006716586.1 | |||
XM_006716524.1 | 150 | Intron | XP_006716587.1 | |||
XM_006716525.1 | 150 | Intron | XP_006716588.1 | |||
XM_011516905.2 | 150 | Intron | XP_011515207.1 | |||
XM_011516906.2 | 150 | Intron | XP_011515208.1 | |||
XM_017013168.1 | 150 | Intron | XP_016868657.1 | |||
XM_017013169.1 | 150 | Intron | XP_016868658.1 | |||
XM_017013170.1 | 150 | Intron | XP_016868659.1 | |||
XM_017013171.1 | 150 | Intron | XP_016868660.1 | |||
XM_017013172.1 | 150 | Intron | XP_016868661.1 | |||
XM_017013173.1 | 150 | Intron | XP_016868662.1 |
PYCRL - pyrroline-5-carboxylate reductase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIGD5 - tigger transposable element derived 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032862.4 | 150 | Missense Mutation | ATG,ATT | M,I 50 | NP_116251.4 |