Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGCGGCTGATGAGGAGGGCGCAG[A/C]GGGTTGGAGCTCGGGGAGGCCCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612641 MIM: 610772 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANK1 - ankyrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NKX6-3 - NK6 homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152568.3 | 77 | Missense Mutation | NP_689781.1 | |||
XM_017013143.1 | 77 | Intron | XP_016868632.1 |