Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCACCTCCCAAATAGAGTGAGTC[G/T]TCTTCCATGTCGCCCGCAACAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610186 | ||||||||||||||||||||
Literature Links: |
FAM66D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM66D - family with sequence similarity 66 member D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC392196 - ubiquitin specific peptidase 17-like family member 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP17L2 - ubiquitin specific peptidase 17-like family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USP17L7 - ubiquitin specific peptidase 17-like family member 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256869.1 | Intron | NP_001243798.1 |