Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615880 MIM: 603780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP39 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP39 - Rho GTPase activating protein 39 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C8orf82 - chromosome 8 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC14 - leucine rich repeat containing 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272036.1 | 1360 | UTR 3 | NP_001258965.1 | |||
NM_014665.3 | 1360 | UTR 3 | NP_055480.1 | |||
XM_005272358.4 | 1360 | UTR 3 | XP_005272415.1 | |||
XM_005272359.4 | 1360 | UTR 3 | XP_005272416.1 | |||
XM_005272360.4 | 1360 | UTR 3 | XP_005272417.1 | |||
XM_017014005.1 | 1360 | UTR 3 | XP_016869494.1 |
LRRC24 - leucine rich repeat containing 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024678.3 | 1360 | Missense Mutation | ACC,GCC | T,A 410 | NP_001019849.2 |
RECQL4 - RecQ like helicase 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |