Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTAATGTTTACCACAAACCCAG[C/T]GAAGCTGATGATCTGTCTGAAGGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614477 | ||||||||||||||||||||
Literature Links: |
C8orf37 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C8orf37 - chromosome 8 open reading frame 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_177965.3 | 1919 | Missense Mutation | CCC,CTC | P,L 201 | NP_808880.1 | |
XM_005250799.3 | 1919 | Missense Mutation | CCC,CTC | P,L 282 | XP_005250856.2 |