Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGTTAGAAATTGTATTCATACCT[C/T]CATCATCATCATCATTTGAAAGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609353 MIM: 611210 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESCO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESCO2 - establishment of sister chromatid cohesion N-acetyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017420.2 | 1209 | Intron | NP_001017420.1 | |||
XM_011544421.2 | 1209 | Intron | XP_011542723.1 | |||
XM_011544422.2 | 1209 | Intron | XP_011542724.1 |
PBK - PDZ binding kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278945.1 | 1209 | Missense Mutation | GAA,GGA | E,G 268 | NP_001265874.1 | |
NM_018492.3 | 1209 | Missense Mutation | GAA,GGA | E,G 257 | NP_060962.2 | |
XM_006716368.2 | 1209 | Missense Mutation | GAA,GGA | E,G 257 | XP_006716431.1 |