Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATTTGGTTATGGTTTTACTTGA[C/T]GGGTGGACGTAAGTTCCACTAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBR5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
UBR5 - ubiquitin protein ligase E3 component n-recognin 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282873.1 | Intron | NP_001269802.1 | ||||
NM_015902.5 | Intron | NP_056986.2 | ||||
XM_005250962.4 | Intron | XP_005251019.1 | ||||
XM_011517104.2 | Intron | XP_011515406.2 | ||||
XM_011517105.2 | Intron | XP_011515407.2 | ||||
XM_011517106.2 | Intron | XP_011515408.2 | ||||
XM_017013533.1 | Intron | XP_016869022.1 | ||||
XM_017013534.1 | Intron | XP_016869023.1 |
UBR5-AS1 - UBR5 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |