Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGGTGGCACCTCTGAGAAAG[A/G]AAGCCTGTAGGCGTTCCTGGAATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST3GAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST3GAL1 - ST3 beta-galactoside alpha-2,3-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003033.3 | Intron | NP_003024.1 | ||||
NM_173344.2 | Intron | NP_775479.1 | ||||
XM_005251025.4 | Intron | XP_005251082.1 | ||||
XM_006716617.1 | Intron | XP_006716680.1 | ||||
XM_011517225.1 | Intron | XP_011515527.1 | ||||
XM_017013736.1 | Intron | XP_016869225.1 | ||||
XM_017013737.1 | Intron | XP_016869226.1 |