Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCATGATTGGCAAGGCAGGTGAG[C/T]AGATTAGGAAATGAGTCTGAAAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114107 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP3CC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP3CC - protein phosphatase 3 catalytic subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243974.1 | Intron | NP_001230903.1 | ||||
NM_001243975.1 | Intron | NP_001230904.1 | ||||
NM_005605.4 | Intron | NP_005596.2 | ||||
XM_017013610.1 | Intron | XP_016869099.1 | ||||
XM_017013611.1 | Intron | XP_016869100.1 |