Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATCCACATATTTGTTTCTATGC[A/G]ATAACCGATATATTTTTGAGTTACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKIA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKIA - protein kinase (cAMP-dependent, catalytic) inhibitor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006823.3 | Intron | NP_006814.1 | ||||
NM_181839.2 | Intron | NP_862822.1 | ||||
XM_017013620.1 | Intron | XP_016869109.1 | ||||
XM_017013621.1 | Intron | XP_016869110.1 | ||||
XM_017013622.1 | Intron | XP_016869111.1 |
PKIA-AS1 - PKIA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |