Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGCCTTGCCTGACCCTGAGCCAG[A/G]GGAGTGGGCCTCAGGGGCTGCTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606663 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOXL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOXL2 - lysyl oxidase like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002318.2 | Intron | NP_002309.1 |
R3HCC1 - R3H domain and coiled-coil containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |