Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTCCCATTACTGGAGAGATCTCT[A/G]GGCTCTATTGAGGCTTTCTTGAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD46 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD46 - ankyrin repeat domain 46 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270377.1 | Intron | NP_001257306.1 | ||||
NM_001270378.1 | Intron | NP_001257307.1 | ||||
NM_001270379.1 | Intron | NP_001257308.1 | ||||
NM_198401.3 | Intron | NP_940683.1 | ||||
XM_006716509.1 | Intron | XP_006716572.1 | ||||
XM_017013106.1 | Intron | XP_016868595.1 |