Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGGCCCTCACCCACAGATGG[C/T]GACTCAGGGCTGTGCTCCCCCCGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608486 MIM: 601445 | ||||||||||||||||||||
Literature Links: |
MTSS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MTSS1 - metastasis suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282971.1 | 2043 | Silent Mutation | TCA,TCG | S,S 651 | NP_001269900.1 | |
NM_001282974.1 | 2043 | Silent Mutation | TCA,TCG | S,S 622 | NP_001269903.1 | |
NM_014751.5 | 2043 | Silent Mutation | TCA,TCG | S,S 647 | NP_055566.3 | |
XM_005251111.1 | 2043 | Silent Mutation | TCA,TCG | S,S 760 | XP_005251168.1 | |
XM_005251113.1 | 2043 | Silent Mutation | TCA,TCG | S,S 695 | XP_005251170.1 | |
XM_005251118.1 | 2043 | Silent Mutation | TCA,TCG | S,S 586 | XP_005251175.1 | |
XM_006716700.1 | 2043 | Silent Mutation | TCA,TCG | S,S 759 | XP_006716763.1 | |
XM_006716701.1 | 2043 | Silent Mutation | TCA,TCG | S,S 756 | XP_006716764.1 | |
XM_006716702.1 | 2043 | Silent Mutation | TCA,TCG | S,S 743 | XP_006716765.1 | |
XM_006716703.1 | 2043 | Silent Mutation | TCA,TCG | S,S 678 | XP_006716766.1 | |
XM_006716704.1 | 2043 | Silent Mutation | TCA,TCG | S,S 650 | XP_006716767.1 | |
XM_006716705.1 | 2043 | Silent Mutation | TCA,TCG | S,S 634 | XP_006716768.1 | |
XM_006716706.1 | 2043 | Silent Mutation | TCA,TCG | S,S 569 | XP_006716769.1 | |
XM_011517403.1 | 2043 | Silent Mutation | TCA,TCG | S,S 693 | XP_011515705.1 | |
XM_011517404.2 | 2043 | Silent Mutation | TCA,TCG | S,S 679 | XP_011515706.1 | |
XM_011517407.1 | 2043 | Silent Mutation | TCA,TCG | S,S 402 | XP_011515709.1 | |
XM_017014086.1 | 2043 | Silent Mutation | TCA,TCG | S,S 755 | XP_016869575.1 | |
XM_017014087.1 | 2043 | Silent Mutation | TCA,TCG | S,S 735 | XP_016869576.1 | |
XM_017014088.1 | 2043 | Silent Mutation | TCA,TCG | S,S 731 | XP_016869577.1 | |
XM_017014089.1 | 2043 | Silent Mutation | TCA,TCG | S,S 730 | XP_016869578.1 | |
XM_017014090.1 | 2043 | Silent Mutation | TCA,TCG | S,S 705 | XP_016869579.1 | |
XM_017014091.1 | 2043 | Silent Mutation | TCA,TCG | S,S 697 | XP_016869580.1 | |
XM_017014092.1 | 2043 | Silent Mutation | TCA,TCG | S,S 691 | XP_016869581.1 | |
XM_017014093.1 | 2043 | Silent Mutation | TCA,TCG | S,S 646 | XP_016869582.1 | |
XM_017014094.1 | 2043 | Silent Mutation | TCA,TCG | S,S 630 | XP_016869583.1 | |
XM_017014095.1 | 2043 | Silent Mutation | TCA,TCG | S,S 626 | XP_016869584.1 | |
XM_017014096.1 | 2043 | Silent Mutation | TCA,TCG | S,S 625 | XP_016869585.1 | |
XM_017014097.1 | 2043 | Silent Mutation | TCA,TCG | S,S 621 | XP_016869586.1 | |
XM_017014098.1 | 2043 | Silent Mutation | TCA,TCG | S,S 582 | XP_016869587.1 | |
XM_017014099.1 | 2043 | Silent Mutation | TCA,TCG | S,S 581 | XP_016869588.1 | |
XM_017014100.1 | 2043 | Silent Mutation | TCA,TCG | S,S 565 | XP_016869589.1 | |
XM_017014101.1 | 2043 | Silent Mutation | TCA,TCG | S,S 541 | XP_016869590.1 |
NDUFB9 - NADH:ubiquinone oxidoreductase subunit B9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |