Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCAGCCCCACCCTGACTCACC[A/G]GGGGCAGTGCTGACAGCTGGTTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604819 MIM: 607733 | ||||||||||||||||||||
Literature Links: |
MIR937 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR937 - microRNA 937 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PUF60 - poly(U) binding splicing factor 60 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCRIB - scribbled planar cell polarity protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015356.4 | 648 | Missense Mutation | CCG,CTG | P,L 214 | NP_056171.3 | |
NM_182706.4 | 648 | Missense Mutation | CCG,CTG | P,L 214 | NP_874365.3 |