Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGTCCCCGGGGCATTCTGGGC[A/G]GGAAGGCCCTCCTCCATCCAGAGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENTPD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENTPD4 - ectonucleoside triphosphate diphosphohydrolase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128930.2 | 2072 | Silent Mutation | CCC,CCT | P,P 600 | NP_001122402.1 | |
NM_004901.4 | 2072 | Silent Mutation | CCC,CCT | P,P 608 | NP_004892.1 |