Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGATAGATGGCAGTTATTCTAGA[C/T]AGCAGTAAATAACCTTGAGCAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LACTB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LACTB2 - lactamase beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XKR9 - XK related 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001011720.1 | Intron | NP_001011720.1 | ||||
NM_001287258.1 | Intron | NP_001274187.1 | ||||
NM_001287259.1 | Intron | NP_001274188.1 | ||||
NM_001287260.1 | Intron | NP_001274189.1 | ||||
XM_006716447.3 | Intron | XP_006716510.1 | ||||
XM_011517524.2 | Intron | XP_011515826.1 | ||||
XM_011517525.2 | Intron | XP_011515827.1 | ||||
XM_011517526.2 | Intron | XP_011515828.1 | ||||
XM_011517527.2 | Intron | XP_011515829.1 | ||||
XM_017013404.1 | Intron | XP_016868893.1 | ||||
XM_017013405.1 | Intron | XP_016868894.1 |