Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGAAAAGGCTATTAAAACGTTCC[C/T]GATTATCCTCAGGTAAATCATCTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613203 MIM: 604912 | ||||||||||||||||||||
Literature Links: |
DSCC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DSCC1 - DNA replication and sister chromatid cohesion 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024094.2 | 1139 | Missense Mutation | CAG,CGG | Q,R 335 | NP_076999.2 | |
XM_005251065.3 | 1139 | Missense Mutation | CAG,CGG | Q,R 290 | XP_005251122.1 |
TAF2 - TATA-box binding protein associated factor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |