Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCTGCCCTGTGGCCTCGCTCCT[C/T]GGAGGGTAAGGCGGCCGGGGGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 222745 | ||||||||||||||||||||
Literature Links: |
DECR1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DECR1 - 2,4-dienoyl-CoA reductase 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001359.1 | 220 | Missense Mutation | CGG,TGG | R,W 22 | NP_001350.1 | |
XM_005250808.4 | 220 | UTR 5 | XP_005250865.1 | |||
XM_011516880.2 | 220 | UTR 5 | XP_011515182.1 | |||
XM_017013147.1 | 220 | UTR 5 | XP_016868636.1 | |||
XM_017013148.1 | 220 | UTR 5 | XP_016868637.1 |