Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCCCAGCAGGCCCACGTTGTGCAG[C/G]AGGAAGAGGAGCCAGGGCCGCGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606027 MIM: 607059 | ||||||||||||||||||||
Literature Links: |
CPSF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPSF1 - cleavage and polyadenylation specific factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC39A4 - solute carrier family 39 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001280557.1 | 1798 | Silent Mutation | CTC,CTG | L,L 124 | NP_001267486.1 | |
NM_017767.2 | 1798 | Silent Mutation | CTC,CTG | L,L 597 | NP_060237.2 | |
NM_130849.3 | 1798 | Silent Mutation | CTC,CTG | L,L 622 | NP_570901.2 | |
XM_006716599.1 | 1798 | Silent Mutation | CTC,CTG | L,L 571 | XP_006716662.1 |