Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTCATGCAGGCGCAGGCCCTGGA[A/G]GAGATGCGGCGCCTAGTCGCCTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613483 MIM: 603711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BHLHE22 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BHLHE22 - basic helix-loop-helix family member e22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152414.4 | 1456 | Silent Mutation | GAA,GAG | E,E 301 | NP_689627.1 |
CYP7B1 - cytochrome P450 family 7 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC401463 - uncharacterized LOC401463 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |