Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGAACCTAAGAAAAGCGAGGGGC[C/T]GAGCGATCGTTTGTCGTGGTCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608486 MIM: 601445 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTSS1 - metastasis suppressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282971.1 | 2352 | Silent Mutation | TCA,TCG | S,S 754 | NP_001269900.1 | |
NM_001282974.1 | 2352 | Silent Mutation | TCA,TCG | S,S 725 | NP_001269903.1 | |
NM_014751.5 | 2352 | Silent Mutation | TCA,TCG | S,S 750 | NP_055566.3 | |
XM_005251111.1 | 2352 | Silent Mutation | TCA,TCG | S,S 863 | XP_005251168.1 | |
XM_005251113.1 | 2352 | Silent Mutation | TCA,TCG | S,S 798 | XP_005251170.1 | |
XM_005251118.1 | 2352 | Silent Mutation | TCA,TCG | S,S 689 | XP_005251175.1 | |
XM_006716700.1 | 2352 | Silent Mutation | TCA,TCG | S,S 862 | XP_006716763.1 | |
XM_006716701.1 | 2352 | Silent Mutation | TCA,TCG | S,S 859 | XP_006716764.1 | |
XM_006716702.1 | 2352 | Silent Mutation | TCA,TCG | S,S 846 | XP_006716765.1 | |
XM_006716703.1 | 2352 | Silent Mutation | TCA,TCG | S,S 781 | XP_006716766.1 | |
XM_006716704.1 | 2352 | Silent Mutation | TCA,TCG | S,S 753 | XP_006716767.1 | |
XM_006716705.1 | 2352 | Silent Mutation | TCA,TCG | S,S 737 | XP_006716768.1 | |
XM_006716706.1 | 2352 | Silent Mutation | TCA,TCG | S,S 672 | XP_006716769.1 | |
XM_011517403.1 | 2352 | Silent Mutation | TCA,TCG | S,S 796 | XP_011515705.1 | |
XM_011517404.2 | 2352 | Silent Mutation | TCA,TCG | S,S 782 | XP_011515706.1 | |
XM_011517407.1 | 2352 | Silent Mutation | TCA,TCG | S,S 505 | XP_011515709.1 | |
XM_017014086.1 | 2352 | Silent Mutation | TCA,TCG | S,S 858 | XP_016869575.1 | |
XM_017014087.1 | 2352 | Silent Mutation | TCA,TCG | S,S 838 | XP_016869576.1 | |
XM_017014088.1 | 2352 | Silent Mutation | TCA,TCG | S,S 834 | XP_016869577.1 | |
XM_017014089.1 | 2352 | Silent Mutation | TCA,TCG | S,S 833 | XP_016869578.1 | |
XM_017014090.1 | 2352 | Silent Mutation | TCA,TCG | S,S 808 | XP_016869579.1 | |
XM_017014091.1 | 2352 | Silent Mutation | TCA,TCG | S,S 800 | XP_016869580.1 | |
XM_017014092.1 | 2352 | Silent Mutation | TCA,TCG | S,S 794 | XP_016869581.1 | |
XM_017014093.1 | 2352 | Silent Mutation | TCA,TCG | S,S 749 | XP_016869582.1 | |
XM_017014094.1 | 2352 | Silent Mutation | TCA,TCG | S,S 733 | XP_016869583.1 | |
XM_017014095.1 | 2352 | Silent Mutation | TCA,TCG | S,S 729 | XP_016869584.1 | |
XM_017014096.1 | 2352 | Silent Mutation | TCA,TCG | S,S 728 | XP_016869585.1 | |
XM_017014097.1 | 2352 | Silent Mutation | TCA,TCG | S,S 724 | XP_016869586.1 | |
XM_017014098.1 | 2352 | Silent Mutation | TCA,TCG | S,S 685 | XP_016869587.1 | |
XM_017014099.1 | 2352 | Silent Mutation | TCA,TCG | S,S 684 | XP_016869588.1 | |
XM_017014100.1 | 2352 | Silent Mutation | TCA,TCG | S,S 668 | XP_016869589.1 | |
XM_017014101.1 | 2352 | Silent Mutation | TCA,TCG | S,S 644 | XP_016869590.1 |
NDUFB9 - NADH:ubiquinone oxidoreductase subunit B9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |