Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACATAAGCCATTGCTGGCCTCCTT[A/G]GAGGACTTAGAGAATCCTGAGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPL23AP53 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPL23AP53 - ribosomal protein L23a pseudogene 53 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF596 - zinc finger protein 596 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042415.2 | Intron | NP_001035880.1 | ||||
NM_001042416.2 | Intron | NP_001035881.1 | ||||
NM_001287254.1 | Intron | NP_001274183.1 | ||||
NM_001287255.1 | Intron | NP_001274184.1 | ||||
NM_001287256.1 | Intron | NP_001274185.1 | ||||
NM_001287399.1 | Intron | NP_001274328.1 | ||||
NM_173539.3 | Intron | NP_775810.2 | ||||
XM_011534744.1 | Intron | XP_011533046.1 | ||||
XM_017013166.1 | Intron | XP_016868655.1 |