Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAATAAGAAACTACTCTTAGCACA[A/G]AAATAACAGAAAATATGCTCACATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 170993 | ||||||||||||||||||||
Literature Links: |
PEX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PEX2 - peroxisomal biogenesis factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000318.2 | 1256 | Silent Mutation | TTC,TTT | F,F 266 | NP_000309.1 | |
NM_001079867.1 | 1256 | Silent Mutation | TTC,TTT | F,F 266 | NP_001073336.1 | |
NM_001172086.1 | 1256 | Silent Mutation | TTC,TTT | F,F 266 | NP_001165557.1 | |
NM_001172087.1 | 1256 | Silent Mutation | TTC,TTT | F,F 266 | NP_001165558.1 |