Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCGCGGATTCCCTGTACCCCACAC[C/T]GTCAGGGCCGAGCCGCTGCCTGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616408 MIM: 137020 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PYCRL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PYCRL - pyrroline-5-carboxylate reductase-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSTA3 - tissue specific transplantation antigen P35B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317783.1 | 745 | Silent Mutation | ACA,ACG | T,T 212 | NP_001304712.1 | |
NM_003313.3 | 745 | Silent Mutation | ACA,ACG | T,T 206 | NP_003304.1 | |
XM_005251051.3 | 745 | Silent Mutation | ACA,ACG | T,T 206 | XP_005251108.2 | |
XM_011517269.1 | 745 | Silent Mutation | ACA,ACG | T,T 206 | XP_011515571.1 |