Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGCCTCCATCTGCTGGGTGTC[A/G]GCCAATATCTTCGGTAAGTTCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 188450 | ||||||||||||||||||||
Literature Links: |
TG PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TG - thyroglobulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003235.4 | 112 | Silent Mutation | TCA,TCG | S,S 18 | NP_003226.4 | |
XM_005251038.4 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_005251095.1 | |
XM_005251040.4 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_005251097.1 | |
XM_005251042.4 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_005251099.1 | |
XM_006716622.3 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_006716685.1 | |
XM_017013793.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869282.1 | |
XM_017013794.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869283.1 | |
XM_017013795.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869284.1 | |
XM_017013796.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869285.1 | |
XM_017013797.1 | 112 | UTR 5 | XP_016869286.1 | |||
XM_017013798.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869287.1 | |
XM_017013799.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869288.1 | |
XM_017013800.1 | 112 | Silent Mutation | TCA,TCG | S,S 18 | XP_016869289.1 |