Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGAACAGAAGGGCACCCACAAC[A/G]TCTGCAGGAATTTCAGGTATCTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610657 MIM: 602019 | ||||||||||||||||||||
Literature Links: |
KIAA0196 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0196 - KIAA0196 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014846.3 | 3897 | Silent Mutation | GAC,GAT | D,D 1121 | NP_055661.3 | |
XM_005251120.3 | 3897 | Silent Mutation | GAC,GAT | D,D 973 | XP_005251177.1 | |
XM_011517409.1 | 3897 | Intron | XP_011515711.1 | |||
XM_017014113.1 | 3897 | Silent Mutation | GAC,GAT | D,D 1121 | XP_016869602.1 |
SQLE - squalene epoxidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |