Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCAGGCTTAGCTTCGGGCGGCAG[C/T]GGTCTGGGGCTGCTCGGACTGGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608861 | ||||||||||||||||||||
Literature Links: |
ATP6V1H PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V1H - ATPase H+ transporting V1 subunit H | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015941.3 | 1642 | Missense Mutation | NP_057025.2 | |||
NM_213619.2 | 1642 | Missense Mutation | NP_998784.1 | |||
NM_213620.2 | 1642 | Missense Mutation | NP_998785.1 | |||
XM_006716455.2 | 1642 | Missense Mutation | XP_006716518.1 | |||
XM_011517542.1 | 1642 | Missense Mutation | XP_011515844.1 |