Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGACCGCCTTTCTCTCTCCGGTGC[C/T]TCCTGGTGGCCGCCGAGGGCATTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 122560 MIM: 606469 | ||||||||||||||||||||
Literature Links: |
CRH PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRH - corticotropin releasing hormone | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000756.3 | 1143 | Silent Mutation | GAA,GAG | E,E 146 | NP_000747.1 | |
XM_017013090.1 | 1143 | Silent Mutation | GAA,GAG | E,E 146 | XP_016868579.1 | |
XM_017013091.1 | 1143 | Silent Mutation | GAA,GAG | E,E 146 | XP_016868580.1 | |
XM_017013092.1 | 1143 | Silent Mutation | GAA,GAG | E,E 146 | XP_016868581.1 | |
XM_017013093.1 | 1143 | Silent Mutation | GAA,GAG | E,E 146 | XP_016868582.1 |
TRIM55 - tripartite motif containing 55 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |