Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGATTTACTTCAAAATGGCTCA[C/T]ACAAACTTTTGGTATAAAAGATAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM65 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM65 - transmembrane protein 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_194291.2 | 4206 | UTR 3 | NP_919267.2 | |||
XM_011516847.2 | 4206 | Intron | XP_011515149.1 | |||
XM_017013104.1 | 4206 | Intron | XP_016868593.1 | |||
XM_017013105.1 | 4206 | Intron | XP_016868594.1 |