Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGGCTCCTGGGCTCGGCGAGGA[C/G]TGCAGGGAGATGAAGGAAGGCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610303 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAFA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAFA - MAF bZIP transcription factor A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZC3H3 - zinc finger CCCH-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015117.2 | 2777 | Missense Mutation | CAC,CAG | H,Q 916 | NP_055932.2 | |
XM_006716536.3 | 2777 | Missense Mutation | CAC,CAG | H,Q 978 | XP_006716599.2 | |
XM_011516943.2 | 2777 | Intron | XP_011515245.1 | |||
XM_011516944.2 | 2777 | Intron | XP_011515246.1 | |||
XM_017013248.1 | 2777 | Missense Mutation | CAC,CAG | H,Q 1044 | XP_016868737.1 | |
XM_017013249.1 | 2777 | Missense Mutation | CAC,CAG | H,Q 1044 | XP_016868738.1 |