Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTCTTTCTGAAACTGACATCACG[A/G]TGCTCCCAGATTTTATGCTGGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 118508 | ||||||||||||||||||||
Literature Links: |
CHRNB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNB3 - cholinergic receptor nicotinic beta 3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000749.3 | 129 | Missense Mutation | ATG,GTG | M,V 1 | NP_000740.1 | |
XM_011544390.1 | 129 | Intron | XP_011542692.1 |