Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGGGTTAGAAAAACGTTTGAT[C/T]CTATAGCTTTCCTCCTCATTAGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601653 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EYA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EYA1 - EYA transcriptional coactivator and phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000503.5 | Intron | NP_000494.2 | ||||
NM_001288574.1 | Intron | NP_001275503.1 | ||||
NM_001288575.1 | Intron | NP_001275504.1 | ||||
NM_172058.3 | Intron | NP_742055.1 | ||||
NM_172059.3 | Intron | NP_742056.1 | ||||
NM_172060.3 | Intron | NP_742057.1 | ||||
XM_011517483.2 | Intron | XP_011515785.1 | ||||
XM_011517484.2 | Intron | XP_011515786.2 | ||||
XM_017013201.1 | Intron | XP_016868690.1 | ||||
XM_017013202.1 | Intron | XP_016868691.1 | ||||
XM_017013203.1 | Intron | XP_016868692.1 | ||||
XM_017013204.1 | Intron | XP_016868693.1 | ||||
XM_017013205.1 | Intron | XP_016868694.1 | ||||
XM_017013206.1 | Intron | XP_016868695.1 | ||||
XM_017013207.1 | Intron | XP_016868696.1 | ||||
XM_017013208.1 | Intron | XP_016868697.1 | ||||
XM_017013209.1 | Intron | XP_016868698.1 | ||||
XM_017013210.1 | Intron | XP_016868699.1 | ||||
XM_017013211.1 | Intron | XP_016868700.1 | ||||
XM_017013212.1 | Intron | XP_016868701.1 | ||||
XM_017013213.1 | Intron | XP_016868702.1 |