Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCAGTCGCACTCTGAGCATCTC[A/G]TCATTGTCGCAGATGATATGGGTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611352 | ||||||||||||||||||||
Literature Links: |
INTS9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
INTS9 - integrator complex subunit 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145159.2 | 1985 | Silent Mutation | GAC,GAT | D,D 619 | NP_001138631.1 | |
NM_001172562.1 | 1985 | Silent Mutation | GAC,GAT | D,D 616 | NP_001166033.1 | |
NM_018250.3 | 1985 | Silent Mutation | GAC,GAT | D,D 640 | NP_060720.2 | |
XM_006716356.2 | 1985 | Silent Mutation | GAC,GAT | D,D 658 | XP_006716419.1 | |
XM_006716357.3 | 1985 | Intron | XP_006716420.1 | |||
XM_011544573.1 | 1985 | Silent Mutation | GAC,GAT | D,D 637 | XP_011542875.1 | |
XM_011544574.1 | 1985 | Silent Mutation | GAC,GAT | D,D 634 | XP_011542876.1 | |
XM_011544575.2 | 1985 | Silent Mutation | GAC,GAT | D,D 634 | XP_011542877.1 | |
XM_011544576.1 | 1985 | Silent Mutation | GAC,GAT | D,D 613 | XP_011542878.1 |