Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCAGCCCGTCCAGCTGGATAGAG[C/T]GCACACAGGCCTCCAGCTGGGCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 130592 MIM: 611552 | ||||||||||||||||||||
Literature Links: |
EEF1D PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EEF1D - eukaryotic translation elongation factor 1 delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005250820.3 | 7085 | Missense Mutation | CAC,CGC | H,R 626 | XP_005250877.1 | |
XM_005250823.1 | 7085 | Intron | XP_005250880.1 | |||
XM_006716519.3 | 7085 | Missense Mutation | CAC,CGC | H,R 625 | XP_006716582.1 | |
XM_006716520.3 | 7085 | Missense Mutation | CAC,CGC | H,R 602 | XP_006716583.1 | |
XM_006716522.1 | 7085 | Missense Mutation | CAC,CGC | H,R 585 | XP_006716585.1 | |
XM_006716523.1 | 7085 | Missense Mutation | CAC,CGC | H,R 585 | XP_006716586.1 | |
XM_006716524.1 | 7085 | Missense Mutation | CAC,CGC | H,R 585 | XP_006716587.1 | |
XM_006716525.1 | 7085 | Missense Mutation | CAC,CGC | H,R 219 | XP_006716588.1 | |
XM_011516905.2 | 7085 | Intron | XP_011515207.1 | |||
XM_011516906.2 | 7085 | Intron | XP_011515208.1 | |||
XM_017013168.1 | 7085 | Intron | XP_016868657.1 | |||
XM_017013169.1 | 7085 | Intron | XP_016868658.1 | |||
XM_017013170.1 | 7085 | Intron | XP_016868659.1 | |||
XM_017013171.1 | 7085 | Intron | XP_016868660.1 | |||
XM_017013172.1 | 7085 | Missense Mutation | CAC,CGC | H,R 561 | XP_016868661.1 | |
XM_017013173.1 | 7085 | Intron | XP_016868662.1 |
MROH6 - maestro heat like repeat family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAPRT - nicotinate phosphoribosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286829.1 | 7085 | Intron | NP_001273758.1 | |||
NM_145201.5 | 7085 | Intron | NP_660202.3 | |||
XM_017013975.1 | 7085 | Intron | XP_016869464.1 | |||
XM_017013976.1 | 7085 | Intron | XP_016869465.1 | |||
XM_017013977.1 | 7085 | Intron | XP_016869466.1 | |||
XM_017013978.1 | 7085 | Intron | XP_016869467.1 | |||
XM_017013979.1 | 7085 | Intron | XP_016869468.1 | |||
XM_017013980.1 | 7085 | Intron | XP_016869469.1 |