Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACGAGAAGGCTCAGGTGCAGCTG[A/G]ACTCGGACCACCTGGAGGAAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 162250 | ||||||||||||||||||||
Literature Links: |
NEFM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NEFM - neurofilament, medium polypeptide | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105541.1 | 553 | Intron | NP_001099011.1 | |||
NM_005382.2 | 553 | Missense Mutation | AAC,GAC | N,D 174 | NP_005373.2 |