Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATATATTATCAAAATTAACTTC[A/G]CCTATTCTTTGATTTAAAAATATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM110B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM110B - family with sequence similarity 110 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147189.2 | Intron | NP_671722.1 | ||||
XM_005251324.2 | Intron | XP_005251381.1 | ||||
XM_005251325.3 | Intron | XP_005251382.1 | ||||
XM_005251326.2 | Intron | XP_005251383.1 | ||||
XM_017013948.1 | Intron | XP_016869437.1 |